missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Mitex™ PTFE Membrane Filters

Unlaminated for extreme chemical or temperature conditions

Manufacturer:  EMD Millipore LSWG02500

 View more versions of this product

Catalog No. LSWG02500


 

Specifications

Description

Filters, Membrane; Millipore Mitex; PTFE; 5.0um Pore Size; White; Type: Grid; Diameter: 25mm; Membranes have no backing; Autoclavable; Mean thickness is 125um; Pack of 100

  • Membranes have no backing
  • Mean thickness is 125μm
  • Autoclavable at 121°C (250°F)

Applications:

Clarification of acids, bases and cryogenic fluids; clarification of propellants; analysis of hydraulic fluids; RNA isolation.

Description & Specifications

Specifications

5μm
25mm
100/pk.
0.9psi
125μm
Polytetrafluoroethylene (PTFE)
White
Yes
Clarification of acids, bases, and cryogenic fluids; Clarification of propellants; Analysis of hydraulic fluids; RNA isolation.
Gridded