missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Millistak+™ HC Pod Disposable Depth Filter System

Combine two distinct technologies to enhance filter capacity and retention

Manufacturer:  EMD Millipore™ MD0HC027H1

 View more versions of this product

Catalog No. MD0HC027H1



Description

Description

  • Filter material: cellulose and inorganic filter aid
  • Configurations: 0.11, 0.55, or 1.1 m2 filtration area
  • Low hold-up volume for greater product yield
  • Broad range of 23 media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000L or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Disposable Pod system protects operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Ideal for Primary and Secondary Clarification in Lab, Pilot and Process-Scale Applications

Specifications

Specifications

Each
0.25 in. hose
Yes
0.027m2
Primary clarification directly out of the bioreactor, laboratory filtration
14cm
8.5 in.
Cellulose
Glass Fiber Reinforced Polypropylene
Millistak+™ Lab Scale Pod Filter
D0HC
960 Lmh/Psid
5.3 in.
50psi at 25°C, 15psi at 80°C
22cm
Documents