Please login to your online account to display your discounted pricing

EMD Millipore™ Millipak™ Disposable Filter Units


Removes particles and microorganisms from liquids and gases with pump-driven filter. Merck Millipore™ Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Manufacturer: emd millipore™  MPGL04GK2

 View more versions of this product

Catalog No. MPGL04GK2

  • / Each
 

Description & Specifications

Specifications

Connectors 0.25 in./6.35mm NPTM inlet, 0.25 in./6.35mm stepped hose barb with bell outlet
Product Type Filter unit
Disposable Yes
Quantity (No. per Case) 2 Pack
Diameter (Metric) Outer 7.6cm
Material Polycarbonate
Diameter (English) Outer 3 in.
Filter Type Durapore™
Filtration Area 250cm2
Flow Rate 2.8L/min. at 1.75 bar
For Use With (Application) Laboratory filtration
Sterile Yes
Inlet Pressure 75psi at 25°C
Length (English) 3.5 in.
Length (Metric) 8.9cm
Material (Membrane) PES
Pore Size (Metric) 0.22μm

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity