Please login to your online account to display your discounted pricing

EMD Millipore™ Millipak™ Disposable Filter Units


Removes particles and microorganisms from liquids and gases with pump-driven filter. Merck Millipore™ Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Manufacturer: emd millipore™  MPVL02GH2

 View more versions of this product

Catalog No. MPVL02GH2

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Product Type Filter unit
Connectors 0.25 in./6.35mm stepped hose barb inlet and outlet with bell
Quantity (No. per Case) 2 Pack
Disposable Yes
Flow Rate 0.7L/min. at 1.75 bar
For Use With (Application) Removal of particles and microorganisms from liquids and gases
Filtration Area 100cm2
Pore Size (Metric) 0.1μm
Length (Metric) 8.4cm
Material Polycarbonate
Filter Type Durapore™
Volume (Metric) 10L
Diameter (English) Outer 3 in.
Inlet Pressure 75psi at 25°C
Length (English) 3.3 in.
Material (Membrane) PVDF
Sterile Yes
Diameter (Metric) Outer 7.6cm

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity