missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Millipak™ Disposable Filter Units

Removes particles and microorganisms from liquids and gases with pump-driven filter. Merck Millipore™ Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Manufacturer:  EMD Millipore™ MPSL06CB1

 View more versions of this product

Catalog No. MPSL06CB1


 

Specifications

Description

Filter unit, Disposable; EMD Millipore; Millipak 60 Filling Machine; 300cm2 Filter area; Pore Size: 5μm; 6mm NPTM/6mm Hose Barb

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity

Description & Specifications

Specifications

7.6cm
5μm
Polycarbonate
Filter unit
Yes
300cm2
Removal of particles and microorganisms from liquids and gases
3.7 in.
PVDF
No
1 Pack
0.25 in./6.35mm stepped hose barb inlet and outlet with bell
3 in.
Durapore™
7.6L/min. at 700mbar
75psi at 25°C
9.4cm
30L