missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Millipak™-20 Filter Unit
Description
Non-sterile filter removes 0.22μm particles at point-of-use prior to dispensing
- Standard point-of-use filter
- Nonsterile
Specifications
Specifications
| Pore Size | 0.22 μm |
| Purification Method | Membrane Filter |
| For Use With (Equipment) | Direct-Q™ 3, Milli-Q™ academic, biocel, element, gradient |
| Type | Millipak™-20 Filter Unit |