Please login to your online account to display your discounted pricing

EMD Millipore Millicell™ Culture Plate Inserts


Designed for improved cell differentiation and expanded applications involving analytical cell biology. EMD Millipore Millicell™ Culture Plate Inserts incorporate PTFE Biopore™ membranes with 0.4μm pores. Ready-to-use 30 or 12mm inserts for 6 and 24-well plates.

Manufacturer: emd millipore  PICM01250

 View more versions of this product

Catalog No. PICM01250

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Material Biopore™ PTFE
Culture Area 0.6cm2
Pore Size 0.4μm
Sterile Yes
For Use With (Equipment) 24 Well Plate
Height (Metric) 10.5mm
Diameter (Metric) Filter 12mm

Microporous, transparent membranes

  • Allow access to cell monolayer from both apical and basolateral domains
  • Promote enhanced cell attachment, growth, and differentiation of attachment-dependent and suspension cultures
  • Hydrophilic PTFE Biopore™ membranes for neural growth and embryo development work
  • Inert and allow repeated viewing of living cells through the membrane
  • Suitable for transport and permeability studies, co-culture for cell-cell interaction studies, in vitro toxicity testing, tumor cell metastasis studies, and organotypic cultures
Ready-to-use inserts
  • Fit most 6- or 24-well plates and are easily prepared for SEM and TEM techniques
  • Effective growth area for 12mm diameter inserts: 0.6cm2
  • Effective growth area for 30mm diameter inserts: 4.2cm2
  • Compatible with standard and fluorescent stains