Please login to your online account to display your discounted pricing

EMD Millipore Milli-Q™ Ultrapure Water Systems Accessory, Cartridges


Choose from a wide selection of replacement cartridges

Manufacturer: emd millipore  QTUM000EX

 View more versions of this product

Catalog No. QTUM000EX

  • / Each
 

Description & Specifications

Specifications

Purification Method Activated Carbon, Ion Exchange
For Use With (Equipment) Milli-Q Systems
Feedwater Source Elix, RO, or Distilled
Model Quantum EX
Product Type Polishing Cartridge