Please login to your online account to display your discounted pricing

EMD Millipore Milli-Q™ Ultrapure Water Systems Accessory, Cartridges


Choose from a wide selection of replacement cartridges

Manufacturer: emd millipore  CDUFHF05K

 View more versions of this product

Catalog No. CDUFHF05K

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Milli-Q Systems
Purification Method Ultrafiltration
Model Pyrogard™ 5000
Product Type Ultrafiltration Cartridge