Please login to your online account to display your discounted pricing

EMD Millipore Milli-Q™ Ultrapure Water Systems Accessory, Cartridges


Choose from a wide selection of replacement cartridges

Manufacturer: emd millipore  QGARD00VX

 View more versions of this product

Catalog No. QGAR-D00-VX

  • / Each
 

Description & Specifications

Specifications

Product Type Water purification cartridge
Purification Method Ion exchange, activated Carbon
For Use With (Equipment) Milli-Q Systems
Model Q-Gard VX
Feedwater Source Elix™, reverse osmosis, distilled water