MilliporeSigma™ Milli-Q™ Integral Systems
Produce pressurized Type II pure and Type I ultrapure water in a single unit—directly from tap water by choosing a Millipore Milli-Q™ Integral System.
Specifications
Certifications/Compliance ISO 9001 v.2000, ISO 14001
Contaminant Rejection (Microbiological) <1cfu/mL (with Millipak or Biopak point-of-use filters)
Contaminant Rejection (Organic) <5ppb
Contaminant Rejection (Particulate) <1 (particulates >0.22μm with Millipak or Biopak point-of-use filters)
Endotoxin Concentration Less than0.001EU/mL (pyrogen-free)
View More Specs
Requires: Potable tap water source (as per US-EPA, EP and WHO norms)
Products 1
Catalog Number Mfr. No. Daily Output Includes Model Price Quantity  
Catalog Number Mfr. No. Daily Output Includes Model Price Quantity  
ZRXQ003T0
View Documents
MilliporeSigma™
ZRXQ003T0
70L Q-POD™ and E-POD™ remote dispenser units, Built-In resistivity and TOC meter Integral 3 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Combine proven Elix™ and Milli-Q technologies to allow complete control over the quality and volume of water required in the lab. Separate points of delivery (PODs) offer convenience and are adaptable to match final polishing to specific research applications.
Single Unit Production
  • No upstream pretreatment system is necessary
  • Produces up to 360L/day at up to 2L/min. on demand
Total Water Quality Control
  • Resistivity and TOC monitoring allows control over both ionic and organic contaminants that can impact your results
  • TOC measuring allows you to verify that the contaminant removal process is operating within specifications
  • Precise resistivity measuring ensures that ionic contamination of purified water remains at sub-ppb level
Customized Water Quality
  • POD Paks, placed at the outlet of each dispenser, are designed to remove specific types of contaminants
  • Water quality remains consistent, day after day, to your most stringent specifications
Designed for Flexibility
  • Component design allows best use of space; PODs can be wall mounted
  • Q-POD delivers Type I Water E-POD delivers pressurized Type II Water
Convenient Water Delivery
  • Manual delivery from POD unit plunger—with control from drop-to-drop up to high flow
  • Dispenser (sold separately) can be used handheld for glassware washing or similar tasks
When equipped with a 0.2μm filter, the system delivers high purity water suitable for HPLC, LC-MS, MALDI-ToF-MS, IC, ICP, AA and most common analytical laboratory techniques. When an ultrafiltration cartridge (BioPak™) is utilized at point-of-use, the water is suitable for genomic applications (DEPC-equivalent) and cell culture. Water produced following the system's pretreatment step can be used for basic laboratory needs, such as buffer and reagent preparation, microbiology media preparation, histology, and glassware rinsing.
Order Info
Dimensions, Production Unit: 19L × 13.1W × 19.7 in.H (48.4 × 33.2 × 50cm). Height of POD unit: 22.8 in. (57.9cm). Operating Weight, Production Unit: 52.9 to 61.7 lb. (24 to 28kg); POD Unit: 10.36 lb. (4.7kg). For 100/230V 50/60Hz. Accessory filters and cartridges are available online.
Certifications
ISO 9001 v.2000 and ISO 14001
Specifications
Specifications
ISO 9001 v.2000, ISO 14001
<5ppb
Less than0.001EU/mL (pyrogen-free)
Potable tap water
≤30 pure water; ≤5ppb ultrapure water
87psi (6 bar) (Differential and inlet)
19.7 Production unit, 22.8 POD unit in.
0.056μS/cm at 25°C
24 to 28 Production unit, 4.7 POD unitkg
<1cfu/mL (with Millipak or Biopak point-of-use filters)
<1 (particulates >0.22μm with Millipak or Biopak point-of-use filters)
19 x 13.1 in. (48.4 x 33.2cm)
Type 1 and 2
100/230V 50/60Hz
HPLC, LC-MS, MALDI-ToF-MS, IC, ICP, AA
50 Production unit, 57.9 POD unitcm
52.9 to 61.7 Production unit, 10.36 POD unit lb.
SDS