Learn More
MilliporeSigma™ Milli-Q™ Advantage A10 Water Purification Packs
For use with Advantage A10 system
Supplier:  MilliporeSigma™ QGARDT2X1
 View more versions of this product
Catalog No. QGARDT2X1
Only null left
Explore available promotions
Description
Description
For use with Advantage A10 system
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Certifications
ISO 9001 v.2000 and ISO 14001
Compliance
USP 645 suitability requirements
Specifications
Specifications
Deionized
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Q-Gard T2 Purification Pack
Ion Exchange, Activated Carbon, Depth Filtration
Milli-Q™ Advantage 10 water purification system
Product Suggestions
Product Suggestions