missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Milli-Q™ Advantage A10 Ultrapure Water Purification System Accessory, Quantum Polishing Cartridges
Description
Quantum polishing catridge removes ionic and organic contaminants below trace levels to match required water quality requirements
Quantum™ TEX Features
- Contains Ionex resins for trace ion levels
- Feedwater nature: Ultrapure
- Technology: Activated carbon, Ion exchange
- Contains Organex and Ionex resins for trace levels of organics and ions
- Feedwater nature: Ultrapure
- Technology: Activated carbon, Ion exchange
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Certifications
ISO 9001 v.2000 and ISO 14001
Compliance
USP 645 suitability requirements
Specifications
Specifications
| Purification Method | Ion Exchange |
| Resin Type | Ionex |
| For Use With (Application) | Spectrophotometry, spectroscopy, chromatography, and biochemistry |
| For Use With (Equipment) | Milli-Q™ Systems |
| Type | Quantum TIX Cartridge Filter |