missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Milli-Q™ Series Water Purification System Accessory, Millipak™ 40 Express Final Filter

Catalog No. MPGP04001
Change view
Click to view available options
Millipak Express 40 Filter Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MPGP04001 Millipak Express 40 Filter Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPGP04001 Supplier MilliporeSigma™ Supplier No. MPGP04001

Use for the production of bacteria-free purified water

  • Pore size: 0.22μm
  • Non-sterile

Certifications

ISO 9001 v.2000 and ISO 14001

Compliance

USP 645 suitability requirements

Specifications

Pore Size 0.22 μm
Purification Method Membrane Filter
For Use With (Equipment) Academic, Gradient, Biocel, Synthesis, Direct-Q, and Milli-DI Systems
Type Water Purification System Accessory