Please login to your online account to display your discounted pricing

EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane


Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.22, 0.45 and 0.80µm pore sizes; medical-grade filters available.

Manufacturer: emd millipore  SLAA033SB

 View more versions of this product

Catalog No. SLAA033SB

  • / Pack of 250 Each
 

Description & Specifications

Specifications

Volume (Metric) 100mL
Tip Material Modified Acrylic
Applications Use for sterilization/clarification of water, buffers, and salt solutions
Pore Size 0.8μm
Hold up Volume (After Air Purge) <100μL
Certifications/Compliance Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Sterile Yes
Diameter (Metric) 33mm
Pressure Rating 145psi
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 113°F
Temperature (Metric) 45°C
Quantity 250/pk.
Color Blue
Type Syringe Filter
Material Mixed Cellulose Esters (MCE)

  • 100% integrity tested, individually packaged, and sterilized
  • Female Luer-Lok™ inlet and male Luer slip outlet
  • Manufactured in a controlled environment using an automated process
  • Blue color-coded band clearly indicates which membrane is inside
  • 0.22µm filters are sterilizing grade
  • Larger pore sizes are for clarification and prefiltration
  • Include a Certificate of Quality
MF-Millipore™ (MCE) membrane
  • Mixed cellulose ester provides reliable, general purpose filtration
  • Use for sterilization/clarification of water, buffers, and salt solutions
  • EtO sterilized
Compliance
  • Meet USP Class VI Biological tests for plastics
  • All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.