Please login to your online account to display your discounted pricing

EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane


Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.22, 0.45 and 0.80µm pore sizes; medical-grade filters available.

Manufacturer: emd millipore  SLHA033SS

 View more versions of this product

Catalog No. SLHA033SS

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Volume (Metric) 100mL
Hold up Volume (After Air Purge) <100μL
Pore Size 0.45μm
Tip Material Modified Acrylic
Certifications/Compliance Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 113°F
Temperature (Metric) 45°C
Quantity 50/pk.
Sterile Yes
Color Blue
Type Syringe Filter
Diameter (Metric) 33mm
Material Mixed Cellulose Esters (MCE)
Pressure Rating 145psi
Applications Use for sterilization/clarification of water, buffers, and salt solutions

  • 100% integrity tested, individually packaged, and sterilized
  • Female Luer-Lok™ inlet and male Luer slip outlet
  • Manufactured in a controlled environment using an automated process
  • Blue color-coded band clearly indicates which membrane is inside
  • 0.22µm filters are sterilizing grade
  • Larger pore sizes are for clarification and prefiltration
  • Include a Certificate of Quality
MF-Millipore™ (MCE) membrane
  • Mixed cellulose ester provides reliable, general purpose filtration
  • Use for sterilization/clarification of water, buffers, and salt solutions
  • EtO sterilized
Compliance
  • Meet USP Class VI Biological tests for plastics
  • All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.