Please login to your online account to display your discounted pricing

EMD Millipore Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane


Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.1, 0.22, 0.45 and 5.0µm pore sizes; medical-grade filters available.

Manufacturer: emd millipore  SLGV033RB

 View more versions of this product

Catalog No. SLGV033RB

  • / Pack of 250 Each
 

Description & Specifications

Specifications

Hold up Volume (After Air Purge) <100μL
Volume (Metric) <200mL
Certifications/Compliance Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Inlet Style Female Luer Lok
Temperature (English) 113°F
Pore Size 0.22μm
Sterile Yes
Tip Material Modified Acrylic
Diameter (Metric) 33mm
Pressure Rating 145psi
Outlet Style Male Luer Slip
Temperature (Metric) 45°C
Quantity 250/pk.
Color Yellow
Type Syringe Filter
Material Durapore Hydrophilic Polyvinylidene Fluoride (PVDF)
Applications Use for sterilization/clarification of water, buffers, and salt solutions

  • 100% integrity tested, individually packaged, and sterilized
  • Female Luer-Lok™ inlet and male Luer slip outlet
  • Manufactured in a controlled environment using an automated process
  • Yellow color-coded band clearly indicates which membrane is inside
  • ≤0.22µm filters are sterilizing grade
  • Larger pore sizes are for clarification and prefiltration
  • Include a Certificate of Quality

Durapore™ (PVDF) membrane

  • Lowest protein binding sterilization/clarification of biological solutions
  • Made of polyvinylidene fluoride (PVFD)
  • Gamma irradiation sterilized
Compliance
  • Meet USP Class VI Biological tests for plastics
  • All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.