missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0µm pore sizes
$245.00 - $1,040.00
Specifications
Certifications/Compliance | Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S. |
---|---|
Color | Yellow |
Applications | Use for sterilization/clarification of water, buffers, and salt solutions |
Inlet Style | Female Luer Lok |
Material | Durapore Hydrophilic Polyvinylidene Fluoride (PVDF) |
Catalog Number | Mfr. No. | Pore Size | Type | Diameter (Metric) | Tip Material | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Type | Diameter (Metric) | Tip Material | Price | Quantity | ||
SLVV033RS
|
MilliporeSigma™
SLVV033RS |
0.1μm | Syringe Filter | 33mm | Modified Acrylic |
Pack of 50 Each for $262.00
|
|
SLGV013SL
|
MilliporeSigma™
SLGV013SL |
0.22μm | Syringe Filter | 13mm | Polyethylene, Polyester |
Pack of 100 Each for $433.00
|
|
SLGV004SL
|
MilliporeSigma™
SLGV004SL |
0.22μm | Syringe Filter | 4mm | Polyethylene, Polyester |
Pack of 100 Each for $346.00
|
|
SLGV033RB
|
MilliporeSigma™
SLGV033RB |
0.22μm | Syringe Filter | 33mm | Modified Acrylic |
Pack of 250 Each for $1,000.00
|
|
SLGV033RS
|
MilliporeSigma™
SLGV033RS |
0.22μm | Syringe Filter | 33mm | Modified Acrylic |
Pack of 50 Each for $262.00
|
|
SLHV004SL
|
MilliporeSigma™
SLHV004SL |
0.45μm | Syringe Filter | 4mm | Polyethylene, Polyester |
Pack of 100 Each for $346.00
|
|
SLHV013SL
|
MilliporeSigma™
SLHV013SL |
0.45μm | Syringe Filter | 13mm | Polyethylene |
Pack of 100 Each for $468.00
|
|
SLHV033RS
|
MilliporeSigma™
SLHV033RS |
0.45μm | Syringe Filter | 33mm | Modified Acrylic |
Pack of 50 Each for $365.33
|
|
SLHV033RB
|
MilliporeSigma™
SLHV033RB |
0.45μm | Syringe Filter | 33mm | Modified Acrylic |
Pack of 250 Each for $1,040.00
|
|
SLSV025LS
|
MilliporeSigma™
SLSV025LS |
5μm | Syringe Filter | 25mm | PVC |
Pack of 50 Each for $245.00
|
|
Description
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0μm pore sizes
- 100% integrity tested, individually packaged, and sterilized
- Female Luer-Lok™ inlet and male Luer slip outlet
- Manufactured in a controlled environment using an automated process
- Yellow color-coded band clearly indicates which membrane is inside
- ≤0.22µm filters are sterilizing grade
- Larger pore sizes are for clarification and prefiltration
- Include a Certificate of Quality
Durapore™ (PVDF) membrane
- Lowest protein binding sterilization/clarification of biological solutions
- Made of polyvinylidene fluoride (PVDF)
- Gamma irradiation sterilized
- Meet USP Class VI Biological tests for plastics
- All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Compliance
Meet USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Specifications
Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S. | |
Use for sterilization/clarification of water, buffers, and salt solutions | |
Durapore Hydrophilic Polyvinylidene Fluoride (PVDF) | |
45°C |
Yellow | |
Female Luer Lok | |
Male Luer Slip |