missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane

Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.1, 0.22, 0.45 and 5.0µm pore sizes; medical-grade filters available.

Manufacturer:  EMD Millipore SLGV033RS

 View more versions of this product

Catalog No. SLGV033RS


 

Specifications

Description

Filters; Syringe; Millipore; Millex; Sterile; Durapore (PVDF) Membrane, yellow; Modified acrylic housing; Pore size: 0.22um; Dia.: 33mm; Lowest protein binding sterilization/clarification of biological solutions; 50/Pk.

  • 100% integrity tested, individually packaged, and sterilized
  • Female Luer-Lok™ inlet and male Luer slip outlet
  • Manufactured in a controlled environment using an automated process
  • Yellow color-coded band clearly indicates which membrane is inside
  • ≤0.22µm filters are sterilizing grade
  • Larger pore sizes are for clarification and prefiltration
  • Include a Certificate of Quality

Durapore™ (PVDF) membrane

  • Lowest protein binding sterilization/clarification of biological solutions
  • Made of polyvinylidene fluoride (PVFD)
  • Gamma irradiation sterilized
Compliance
  • Meet USP Class VI Biological tests for plastics
  • All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Description & Specifications

Specifications

Syringe Filter
Durapore Hydrophilic Polyvinylidene Fluoride (PVDF)
Use for sterilization/clarification of water, buffers, and salt solutions
<200mL
Female Luer Lok
113°F
50/pk.
Yes
Yellow
33mm
145psi
<100μL
Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Male Luer Slip
45°C
0.22μm
Modified Acrylic
Promotions

Promotions