EMD Millipore Millex™ Nonsterile Syringe Prefilters
Use to prefilter contaminated solutions
$77.00 - $130.00
Specifications
Applications To prefilter contaminated sollutions
Material AP 20 Glass Fiber
Pore Size 2.0μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Housing Material Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Housing Material Diameter (Metric) Price Quantity  
SLAP05010 EMD Millipore
SLAP05010
Polypropylene 50mm Pack of 10 Each for $77.00
SLAP02550 EMD Millipore
SLAP02550
PVC 25mm Pack of 50 Each for $130.00
Specifications
Description
With Type AP 20 glass fiber (2.0μm nominal pore size) in polyvinyl chloride (PVC) or polypropylene (PP) housings.
SLAP 025 50 has female Luer-Lok™ inlet and male slip Luer outlet SLAP 050 10 has stepped hose barb/female slip Luer interior inlet and outlet.
Description & Specifications
Specifications
To prefilter contaminated sollutions
AP 20 Glass Fiber
2.0μm