MilliporeSigma™ Millex™ Nonsterile Syringe Filters with PVC Housing
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer:  MilliporeSigma™ SLSV025NB
 View more versions of this product
Catalog No. SLSV025NB
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
  • With female Luer-Lok™ inlet and male Luer slip outlet
  • Available in a variety of membrane types
MCE Membrane
  • Mixed cellulose ester membrane for sterilization/clarification of water, buffers, and salt solutions
PVDF Membrane
  • Lowest protein binding
  • Use for aqueous and mild organic solutions
Mitex™-SV
  • Compatible with more aggressive organic solutions and high temperature conditions
Specifications
Specifications
25 mm
Syringe Filter
5 μm
250/Pk.
PVDF
Non-sterile