MilliporeSigma™ Millex™ Nonsterile Syringe Filters with PVC Housing
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
$871.00 - $2,870.00
Specifications
Diameter (Metric) 25 mm
Type Syringe Filter
Sterility Non-sterile
View More Specs
Products 6
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Material Pore Size Quantity Price Quantity  
SLAA025NK
View Documents
MilliporeSigma™
SLAA025NK
Mixed Cellulose Ester 0.8 μm 1000/Pk.
Pack of 1000 Each for $2,050.00
SLAA025NB
View Documents
MilliporeSigma™
SLAA025NB
Mixed Cellulose Ester 0.8 μm 250/Pk.
Pack of 250 Each for $871.00
SLGS025NB
View Documents
MilliporeSigma™
SLGS025NB
Mixed Cellulose Ester 0.22 μm 250/Pk.
Pack of 250 Each for $884.00
SLHA02510
View Documents
MilliporeSigma™
SLHA02510
Mixed Cellulose Ester 0.45 μm 1000/Pk.
Pack of 1000 Each for $2,870.00
SLHA025NB
View Documents
MilliporeSigma™
SLHA025NB
Mixed Cellulose Ester 0.45 μm 250/Pk.
Pack of 250 Each for $884.00
SLSV025NB
View Documents
MilliporeSigma™
SLSV025NB
PVDF 5 μm 250/Pk.
Pack of 250 Each for $921.00
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
  • With female Luer-Lok™ inlet and male Luer slip outlet
  • Available in a variety of membrane types
MCE Membrane
  • Mixed cellulose ester membrane for sterilization/clarification of water, buffers, and salt solutions
PVDF Membrane
  • Lowest protein binding
  • Use for aqueous and mild organic solutions
Mitex™-SV
  • Compatible with more aggressive organic solutions and high temperature conditions
Specifications
Specifications
25 mm
Syringe Filter
Non-sterile