EMD Millipore Millex™ Nonsterile 33mm Syringe Filters
Remove particulates, reduce signal-to-noise ratios and maintain flat baselines. EMD Millipore Millex™ Nonsterile 33mm Syringe Filters are constructed of polyethylene housing with Durapore™ membranes for preparing samples for analysis. Available in two pore sizes.
$220.00 - $1,977.00
Specifications
Housing Material Polyethylene
Membrane Durapore™ Hydrophilic Polyvinylidene Fluoride (PVDF)
Filtration Area (Metric) 4.5cm sq.
Diameter (Metric) 33mm
Hold up Volume (After Air Purge) ≤80µL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
SLGV033NB EMD Millipore
SLGV033NB
0.22μm Pack of 250 Each for $777.00
SLGV033NS EMD Millipore
SLGV033NS
0.22μm Pack of 50 Each for $220.00
SLGV033NK EMD Millipore
SLGV033NK
0.22μm Pack of 1000 Each for $1,977.00
SLHV033NB EMD Millipore
SLHV033NB
0.45μm Pack of 250 Each for $777.00
SLHV033NS EMD Millipore
SLHV033NS
0.45μm Pack of 50 Each for $220.00
SLHV033NK EMD Millipore
SLHV033NK
0.45μm Pack of 1000 Each for $1,977.00
Specifications
Description
  • Low-extractable polyethylene housing
  • Includes a 0.22 or 0.45μm membrane filter for fine filtration
  • Female Luer-Lok™ inlet and Male Luer Slip outlet
  • Durapore™ (PVDF) membrane for lowest protein binding
  • Use with aqueous and mild organic solutions
  • For sample preparation of protein-containing solutions prior to chromatography or other instrument analyses
Description & Specifications
Specifications
Polyethylene
4.5cm sq.
≤80µL
Durapore™ Hydrophilic Polyvinylidene Fluoride (PVDF)
33mm