missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Millex™ Syringe Filter, PES, 0.22 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Supplier: MilliporeSigma™ SLGP033NB
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Low protein binding to minimize interaction with your sample and maximize recovery
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Applications:
Buffer Filtration, Clarification of Aqueous Solutions
Specifications
Single-use | |
PES | |
Non-sterile | |
Analytical sample prep | |
Millex™ | |
Syringe and Syringeless Filter | |
33 mm |
Millipore Express PLUS (PES) | |
0.22 μm | |
4.5 cm2 | |
PP | |
250Pack | |
<200 mL | |
33 mm |