Please login to your online account to display your discounted pricing
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Nylon
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer: emd millipore  SLGN033NB
 View more versions of this product
Catalog No. SLGN033NB
  • / Pack of 250 Each
Description & Specifications
Specifications
Quantity 250/Pk.
Type Syringe Filter
Material Nylon (NYL)
Pore Size 0.2μm
Diameter (Metric) 33mm
  • Broad chemical compatibility Choose for filtering aqueous and organic solutions