Please login to your online account to display your discounted pricing
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Nylon
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer: emd millipore  SLHN033NB
 View more versions of this product
Catalog No. SLHN033NB
  • / Pack of 250 Each
Description & Specifications
Specifications
Pore Size 0.45μm
Diameter (Metric) 33mm
Quantity 250/pk.
Type Syringe Filter
Material Nylon (NYL)
  • Broad chemical compatibility Choose for filtering aqueous and organic solutions