Please login to your online account to display your discounted pricing
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophobic PTFE
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer: emd millipore  SLFG025NS
 View more versions of this product
Catalog No. SLFG025NS
  • / Pack of 50 Each
Description & Specifications
Specifications
Quantity 50/Pk.
Type Syringe Filter
Sterile No
Material Polytetrafluoroethylene (PTFE)
Diameter (Metric) 25mm
Pore Size 0.2μm
Housing Material Polyethylene
  • Compatible with more aggressive organic solutions and high temperature conditions