MilliporeSigma™ Millex™ Syringe Filter, Fluoropore™ PTFE, Hydrophobic, 0.45 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
$291.00 - $3350.00
Specifications
Disposable Single-use
Pore Size 0.45 μm
Sterility Non-sterile
For Use With (Application) Analytical sample prep
View More Specs
Products 7
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Filter Outlet Style Quantity Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Filter Outlet Style Quantity Price Quantity  
SLFH025NK
View Documents Promotion Details
MilliporeSigma™
SLFH025NK
100 mL 25 mm Male Luer slip 1000Pack
Pack of 1000 Each for $3,350.00
SLFH025NS
View Documents Promotion Details
MilliporeSigma™
SLFH025NS
100 mL 25 mm Male Luer slip 50Pack
Pack of 50 Each for $387.00
SLFH025NB
View Documents Promotion Details
MilliporeSigma™
SLFH025NB
100 mL 25 mm Male Luer slip 250Pack
Pack of 250 Each for $1,430.00
SLFHR04NL
View Documents Promotion Details
MilliporeSigma™
SLFHR04NL
1 mL 4.0 mm Male Luer slip 100Pack
Each for $291.00
SLFHX13NK
View Documents Promotion Details
MilliporeSigma™
SLFHX13NK
<10 mL 13 mm Male Luer slip 1000Pack
Pack of 1000 Each for $2,580.00
SLFHX13NL
View Documents Promotion Details
MilliporeSigma™
SLFHX13NL
<10 mL 13 mm Male Luer slip 100Pack
Pack of 100 Each for $393.00
SLFHX13TL
View Documents Promotion Details
MilliporeSigma™
SLFHX13TL
<10 mL 13 mm Tube 100Pack
Pack of 100 Each for $423.00
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Overview of Non-Sterile Millex™ Syringe Filters with Hydrophobic Fluoropore™ (PTFE) Membrane:
  • Ideal for filtering samples in organic solvents, and can also be used to prevent back-streaming of atmospheric moisture in venting applications.
  • Broad chemical compatibility and excellent solvent resistance with organic solutions
  • Housing is made from a low-extractable, high density polyethylene (HDPE) or polypropylene.
Applications:
 Fine Particle Removal and Clarification of Organic Solutions, Vent Filtration, Sterilization of Gases
Specifications
Specifications
Single-use
0.45 μm
Non-sterile
Analytical sample prep
SDS
missing translation for 'documents'
missing translation for 'documents'