Please login to your online account to display your discounted pricing
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophobic PTFE
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer: emd millipore  SLFHR04NL
 View more versions of this product
Catalog No. SLFHR04NL
  • / Pack of 100 Each
Description & Specifications
Specifications
Pore Size 0.45μm
Quantity 100/Pk.
Diameter (Metric) 4.0mm
Material Hydrophobic Polytetrafluoroethylene (PTFE)
Sterile No
Type Syringe Filter
Housing Material Polyethylene
  • Compatible with more aggressive organic solutions and high temperature conditions