MilliporeSigma™ Millex™ Syringe Filter, Hydrophilic PTFE, 0.45 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
$271.00 - $3,770.00
Specifications
Disposable | Yes |
---|---|
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Analytical sample prep |
Temperature (Metric) Max | 45°C |
Catalog Number | Mfr. No. | Diameter (Metric) Filter | Outlet Style | Filter Code | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Filter | Outlet Style | Filter Code | Quantity | Price | Quantity | ||
SLCR025NK
|
MilliporeSigma™
SLCR025NK |
25 mm | Male luer slip | LCR | 1000Pack |
Pack of 1000 Each for $3,770.00
|
|
SLLHR04NL
|
MilliporeSigma™
SLLHR04NL |
4.0 mm | Male Luer slip, stepped | LH | 100Pack |
Each for $271.00
|
|
SLLHR04NK
|
MilliporeSigma™
SLLHR04NK |
4.0 mm | Male Luer slip, stepped | LH | 1000Pack |
Pack of 1000 Each for $2,610.00
|
|
SLLHC13NL
|
MilliporeSigma™
SLLHC13NL |
13 mm | Male Luer slip | LH | 100Pack |
Pack of 100 Each for $361.00
|
|
SLLHH13NK
|
MilliporeSigma™
SLLHH13NK |
13 mm | Male Luer slip | LH | 1000Pack |
Each for $2,910.00
|
|
SLCR013NL
|
MilliporeSigma™
SLCR013NL |
13 mm | Male Luer slip | LCR | 100Pack |
Pack of 100 Each for $421.00
|
|
SLCRT13NL
|
MilliporeSigma™
SLCRT13NL |
13 mm | Tube outlet | LCR | 100Pack |
Pack of 100 Each for $465.00
|
|
SLCR013NK
|
MilliporeSigma™
SLCR013NK |
13 mm | Male Luer slip | LCR | 1000Pack |
Pack of 1000 Each for $2,910.00
|
|
SLLHC25NS
|
MilliporeSigma™
SLLHC25NS |
25 mm | Male Luer slip | LH | 50Pack |
Pack of 50 Each for $299.00
|
|
SLCR033NB
|
MilliporeSigma™
SLCR033NB |
33 mm | Male Luer slip | LCR | 250 |
Pack of 250 Each for $1,070.01
|
|
Description
Overview of Non-Sterile IC Millex™ Syringe Filters:- Ion chromatography certified
- Low level of IC extractables, optimizing background levels in sensitive IC analyses
- Compatible with both aqueous and organic solutions, offering broad chemical compatibility and flexibility in the lab
Specifications
Yes | |
Non-sterile | |
45°C |
0.45 μm | |
Analytical sample prep |