Please login to your online account to display your discounted pricing
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophilic PTFE
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer: emd millipore  SLLHR04NL
 View more versions of this product
Catalog No. SLLHR04NL
  • / Pack of 100 Each
Description & Specifications
Specifications
Housing Material Polyethylene
Pore Size 0.45μm
Quantity 100/Pk.
Diameter (Metric) 4.0mm
Material Polytetrafluoroethylene (PTFE)
Applications For filtering aqueous and organic solutions
Type Syringe Filter