MilliporeSigma™ Millex™ Filters: Inlet and Outlet for Silicone Tubing
Protect vacuum sources from contamination and moisture damage
$186.00 - $1,250.00
Specifications
Autoclavable Yes
Diameter (Metric) 50 mm
Membrane PTFE
For Use With Protect vacuum sources from contamination and moisture damage; Sterilize ambient air entering fermenters; Filter incubator gas to protect against contamination; Filter nitrogen and air used to dry liquid samples
Housing Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
SLFG85000
View Documents
MilliporeSigma™
SLFG85000
0.2μm 100/Pk.
Pack of 100 Each for $1,250.00
SLFG85010
View Documents
MilliporeSigma™
SLFG85010
0.2μm 10/Pk.
Pack of 10 Each for $186.00
Description
Description
Protect vacuum sources from contamination and moisture damage
Stepped hose barb with female Luer slip interior.
  • With hydrophobic PTFE membranes
  • Polypropylene housing (except for SLFG-025-LS and -50, which have PVC housing)
  • Variety of inlet/outlet combinations, including stepped hose barb and 1/8 NPT
  • Nonsterile, autoclavable, bidirectional
Applications:
  • Protect vacuum sources from contamination and moisture damage
  • Sterilize ambient air entering fermenters
  • Filter incubator gas to protect against contamination
  • Filter nitrogen and air used to dry liquid samples
Specifications
Specifications
Yes
PTFE
Polypropylene
50 mm
Protect vacuum sources from contamination and moisture damage; Sterilize ambient air entering fermenters; Filter incubator gas to protect against contamination; Filter nitrogen and air used to dry liquid samples
Non-sterile
SDS