Please login to your online account to display your discounted pricing

EMD Millipore Millex™ Filters: Inlet, Female Luer-Lok; Outlet, Male Luer Slip


Protect vacuum sources from contamination and moisture damage

Manufacturer: emd millipore  SLFG02550

 View more versions of this product

Catalog No. SLFG02550

  • / Pack of 50 Each
 

Description & Specifications

Specifications

For Use With (Application) Protect vacuum sources from contamination and moisture damage; Sterilize ambient air entering fermenters; Filter incubator gas to protect against contamination; Filter nitrogen and air used to dry liquid samples
Pore Size 0.22μm
Material (Membrane) PTFE
Housing Material Polyvinyl Chloride
Sterile No
Autoclavable Yes
Diameter (Metric) Outer 25mm