Please login to your online account to display your discounted pricing
EMD Millipore Microfil™ V Filtration Device
Faster and easier than conventional filter holders used to test drinking water
Manufacturer: emd millipore  MVHABG124
 View more versions of this product
Catalog No. MVHABG124
  • / Pack of 24 Each
Description & Specifications
Specifications
Diameter (Metric) Inner 47mm
Pore Size 0.45μm
Description Fitration Device
Material MCE, Black, gridded