EMD Millipore™ Microfil™ S Filtration Devices
Fast, dependable alternative to conventional multi-step methods
$174.00 - $259.00
Specifications
For Use With (Equipment) EMD Millipore Microfil S Filtration Devices
View More Specs
Products 3
Catalog Number Mfr. No. Material Pore Size Price Quantity  
Catalog Number Mfr. No. Material Pore Size Price Quantity  
MVGSWGS24 View Documents EMD Millipore™
MVGSWGS24
MCE 0.22μm Pack of 24 Each for $251.00
MVHAWGS24 View Documents EMD Millipore™
MVHAWGS24
MCE 0.45μm Pack of 24 Each for $174.00
MVHVWPS24 View Documents EMD Millipore™
MVHVWPS24
PVDF 0.45μm Pack of 24 Each for $259.00
Description
Description
Devices consist of a funnel, protective lid, and filtration support so routine microbiological testing is fast and easy.
  • Ensure consistent recovery of microorganisms
  • Smooth, hydrophobic funnel surface repels aqueous sample residues to ensure that microorganisms are collected on the membrane, not on funnel walls
  • Convex support shape eliminates pleating and distortion of membrane due to expansion
  • Three membrane selections offer versatility for your specific application
Benefits
  • Presterilized, individually packaged No membrane handling prior to filtering
  • No filter holders, funnels and attachments to assemble and sterilize
  • No sterilizing the filter support after each filtration Protective lid reduces external contamination risk Pre-assembled to speed sampling and shorten preparation Easy-to-remove funnel and membrane reduce test time
Compliance
Comply with USP/EP Guidelines. Certificate of Quality demonstrates that the device has been quality-control tested with Pharmacopoeia organisms and demonstrates acceptable growth.
Specifications
Specifications
EMD Millipore Microfil S Filtration Devices