EMD Millipore Microfil™ Filtration System
Convenient filtration system uses presterilized funnels and membranes on lever-action filtration support
$868.00 - $868.00
Includes: 47mm gridded MCE membrane filters or EZ-Pak™ membrane cartridge.
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
MIACFD101 EMD Millipore
MIACFD101
Each for $868.00
Specifications
Description
  • Disposable, stackable, recyclable polypropylene 100mL or 250mL funnels
  • Eliminates autoclaving and minimizes waste
  • Manifold-mounted supports for laboratory use or stand-alone supports for onsite filtration
  • “Push-fit” funnels seal tightly to manifold supports without clamps or O-rings
  • Leak-free operation and uniform microorganism recoveries
  • Faster than all-glass filter apparatus
  • Lever breaks vacuum and lifts membrane for removal with forceps
Order Info
47mm gridded MCE membrane filters or EZ-Pak™ membrane cartridge
Compliance
CEN, Standard Methods for the Examination of Water and Wastewater, WHO guidelines