missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Microcon™ Centrifugal Filters for Protein and DNA Concentration: Ultracel-10 Membrane
Provides efficient concentration, desalting, or buffer exchange of aqueous biological samples.
Manufacturer: MilliporeSigma™ MRCPRT010
Description
- The Microcon centrifugal filters simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes
- The DNA fast flow filter is optimized for the concentration and recovery of genomic DNA with an SDS buffer. The low non-specific binding characteristics of the membrane and the other device components, coupled with its medical-grade o-ring seal, allow the device to accommodate several wash steps with minimal sample loss
Microcon Centrifugal Filters
- Typical recoveries of >95%, even for dilute solutions
- Reverse spin to maximize recovery, even in the smallest samples
- Convenient storage of filtrate or concentrated sample in standard microfuge tube
- Concentration factors up to 100X
- High recovery for small volumes with reverse spin (concentration factor <20X)
- Low-binding Ultracel™ membrane
- Fast processing
Specifications
Non-sterile | |
Ultracel-10 membrane | |
4.5cm |
0.5mL | |
Green |