MilliporeSigma™ Microcon™ Centrifugal Filters for Protein and DNA Concentration: Regenerated Cellulose Membrane
ideal for DNA concentration and desalting applications.
$561.00 - $561.00
Specifications
Membrane Material Regenerated Cellulose
Length (English) 1.8 in.
Length (Metric) 4.5cm
View More Specs
Products 2
Catalog Number Mfr. No. Color MWCO Price Quantity  
Catalog Number Mfr. No. Color MWCO Price Quantity  
MRCF0R030
View Documents
MilliporeSigma™
MRCF0R030
Clear 30 kDa
Pack of 100 Each for $561.00
MRCF0R100
View Documents
MilliporeSigma™
MRCF0R100
Blue 100 kDa
Pack of 100 Each for $561.00
Description
Description
ideal for DNA concentration and desalting applications.
The dual cycle Ethylene Oxide (EtO) treatment renders contaminating DNA non-amplifiable. Available in a pack of 20.
Specifications
Specifications
Regenerated Cellulose
1.8 in.
4.5cm
SDS