missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Microbial Count Samplers

Sterilized, individually packaged, ready-to-use dip testers for Total Count and HPC Total Count

$169.00 - $169.00

Specifications

Color Red
For Use With (Application) Clinical lab, environmental monitoring, food and beverage, water monitoring
Material Styrene Acrylonitrile
Pore Size 0.45μm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
MHPC10025 EMD Millipore
MHPC10025
Pack of 25 Each for $169.00
Specifications

Description

  • Convenient alternative to Standard Methods MF method for routine monitoring of microbiological organisms
  • Consist of plastic paddle with air vent, filter in contact with nutrient-containing pad, and plastic sampling/incubation case
  • 0.45μ filter is marked with a grid to aid in counting colonies on its surface
  • Incubation time: 48 to 72 hours at 25° to 35°C (77° to 95°F)

Magnification of colonies is recommended.

Description & Specifications

Specifications

Red
Clinical lab, environmental monitoring, food and beverage, water monitoring
Styrene Acrylonitrile
0.45μm