Please login to your online account to display your discounted pricing

EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes: 8.0μm Pore Size


For use in Air monitoring, Fluid monitoring, particle collection and analysis, and QC of fluid holding tanks

Manufacturer: emd millipore  SCWP09025

 View more versions of this product

Catalog No. SCWP09025

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Pore Size 8μm
For Use With (Application) For use in Air monitoring; Fluid monitoring; Particle collection and analysis; QC of fluid holding tanks.
Wettability Hydrophilic
Material Mixed Cellulose Esters (MCE)
Diameter (Metric) Outer 90mm
Color White
Accessories

Accessories for Catalog Number: SCWP09025

Item Mfr. No. Item Description Price Per Unit Quantity