Please login to your online account to display your discounted pricing

EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes: 1.2μm Pore Size


Manufacturer: emd millipore  RAWG04700

 View more versions of this product

Catalog No. RAWG04700

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Pore Size 1.2μm
Wettability Hydrophilic
Quantity (No. per Case) 100/Pk.
Material Mixed Cellulose Esters (MCE)
Membrane Type Gridded
Diameter (Metric) Outer 47mm
Color White
Accessories

Accessories for Catalog Number: RAWG04700

Item Mfr. No. Item Description Price Per Unit Quantity