Please login to your online account to display your discounted pricing

EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes: 0.80μm Pore Size


Designed for air monitoring, bioassays, dairy microbiology, particle monitoring and particle removal. EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes, 0.80µm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.

Manufacturer: emd millipore  AAWG01300

 View more versions of this product

Catalog No. AAWG01300

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Bubble Point ≥1.0 bar, air with water
Wettability Hydrophilic
Thickness 150µm
Liquid Flow Rate 190mL/min/cm sq.
Gas Flow Rate 16L/min/cm sq.
For Use With (Application) For air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
Quantity (No. per Case) 100/pk.
Material Mixed Cellulose Esters (MCE)
Pore Size 0.8μm
Membrane Type Gridded
Diameter (Metric) Outer 13mm
Color White

  • Hydrophilic, 150µm thick membranes with a plain or gridded surface
  • Composed of a biologically inert mixture of cellulose acetate and cellulose nitrate
  • Widely used with fuel and aviation turbine fluid contamination SDI, particles and insoluble contaminants
  • White membranes
    Ideal for air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
  • Black membranes
    Ideal for Air monitoring, fluorescent assays, and particle monitoring