missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes: 0.80μm Pore Size

Designed for air monitoring, bioassays, dairy microbiology, particle monitoring and particle removal. EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes, 0.80µm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.

Manufacturer:  EMD Millipore™ AABG04700

 View more versions of this product

Catalog No. AABG04700


 

Description

Description

  • Hydrophilic, 150µm thick membranes with a plain or gridded surface
  • Composed of a biologically inert mixture of cellulose acetate and cellulose nitrate
  • Widely used with fuel and aviation turbine fluid contamination SDI, particles and insoluble contaminants
  • White membranes
    Ideal for air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
  • Black membranes
    Ideal for Air monitoring, fluorescent assays, and particle monitoring
Specifications

Specifications

0.8μm
47mm
100/pk.
For air monitoring, fluorescent assays, and particle monitoring
190mL/min/cm sq.
Hydrophilic
Mixed Cellulose Esters (MCE)
Black
≥1.0 bar, air with water
16L/min/cm sq.
150µm
Gridded
Documents
Accessories