Please login to your online account to display your discounted pricing

EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes: 0.025μm Pore Size


Manufacturer: emd millipore  VSWP09025

 View more versions of this product

Catalog No. VSWP09025

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Material Mixed Cellulose Esters (MCE) (Nitrate and Acetate)
Diameter (Metric) Outer 90mm
Thickness 105μm
Pore Size 0.03μm