missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Membrane Filter Refills and Alternatives

For 37mm monitor

$180.00 - $403.00

Specifications

Diameter (Metric) Outside 3.7cm
Filter Surface Plain
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Filter Code Hydrophobicity Material Pore Size Price Quantity  
Catalog Number Mfr. No. Filter Code Hydrophobicity Material Pore Size Price Quantity  
HAWP03700 EMD Millipore
HAWP03700
HAWP Hydrophilic Mixed Cellulose Ester 0.22μm Pack of 100 Each for $180.00
PVC503700 EMD Millipore
PVC503700
PVC5 Hydrophobic PVC 5μm Pack of 100 Each for $403.00
Specifications

Description

AAWP037PM

  • Matched-weight, plain
  • 0.8μm
  • Thick absorbent pads for liquid monitoring
HAWP03700
  • Gridded
  • 0.45μm
  • Thin absorbent pads for air monitoring

Description & Specifications

Specifications

3.7cm
Plain