EMD Millipore™ MColortest™ Ozone Test Kits
to measure the content of ozone in drinking water, wastewater, process water and disinfecting solutions.
$516.20 - $835.57
Specifications
Product Type Ozone Test Kit
View More Specs
Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
M1187550001 View Documents EMD Millipore™
1187550001
Each for $516.20
M1187580001 View Documents EMD Millipore™
1187580001
Each for $835.57
Description
Description
The test measures the content of ozone in drinking water, wastewater, process water and disinfecting solutions. The concentration can be measured semiquantitatively by using the colour comparator. Method: colorimetric, DPD, with color-disk comparator 0.15 - 0.35 - 0.5 - 0.7 - 1.4 - 2.7 - 5.0 - 7.0 - 10mg/l O₃ MColortest™
Specifications
Specifications
Ozone Test Kit