MilliporeSigma™ MQuant™ pH Indicator Papers
Measure pH onsite without instruments or extensive preparation
Supplier:  MilliporeSigma™ 1095550003
 View more versions of this product
Catalog No. M1095550003
Description
Description
Specifications
Specifications
3.8 to 5.4
<3 to 5.4
10°C to 25°C
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
3 x 4.8 Rolls
Special pH Indicator Paper