MilliporeSigma™ MQuant™ pH Indicator Papers
Measure pH onsite without instruments or extensive preparation
$102.00 - $154.21
Specifications
For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
Quantity 3 x 4.8 Rolls
Storage Requirements 10°C to 25°C
Type Special pH Indicator Paper
View More Specs
Products 3
Catalog Number Mfr. No. pH Range Graduations Price Quantity  
Catalog Number Mfr. No. pH Range Graduations Price Quantity  
M1095560003
View Documents Promotion Details
MilliporeSigma™
1095560003
5.4 to 7 <5.4 to >7 N/A
M1095570003
View Documents Promotion Details
MilliporeSigma™
1095570003
6.4 to 8 6.4 to >8 N/A
M1095550003
View Documents Promotion Details
MilliporeSigma™
1095550003
3.8 to 5.4 <3 to 5.4 N/A
Description
Description
Specifications
Specifications
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
3 x 4.8 Rolls
10°C to 25°C
Special pH Indicator Paper
SDS
missing translation for 'documents'
missing translation for 'documents'