MilliporeSigma™ MColorpHast™ pH Indicator Papers
Measure pH onsite without instruments or extensive preparation
Manufacturer:  MilliporeSigma™ 1095580003
 View more versions of this product
Catalog No. M1095580003
Specifications
Specifications
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
3 x 4.8 Rolls
10° to 25°C
8.2 to 10
<8.2 to 10
8.2 to 10.0 pH
Special pH Indicator Paper
Documents