Please login to your online account to display your discounted pricing
EMD Millipore M Air T™ Agar Cassettes
Ready-to-use cassettes, prefilled with gamma-sterilized medium
Manufacturer: emd millipore  ATSMTTD60
 View more versions of this product
Catalog No. ATSMTTD60
  • / Pack of 60 Each
Description & Specifications
Specifications
For Use With (Equipment) M air T air testing system
For Use With (Application) For the total count of airborne microorganisms in pharmaceutical clean rooms, hospitals, food and beverage applications
Certifications/Compliance USP
Product Type M air T Cassettes
Includes Trypticase Soy Agar media
  • Fast and reliable results with maximum recovery
  • Large agar amount and unique micro-perforated sieve allow air sampling of up to 1 cubic meter
  • Unit-to-unit consistency of agar surface shape
  • Double-sleeve packaged
  • Reproducible results